Assay Details
Target Gene Details
Entrez Gene ID: | 54810 |
Gene Name: | GIPC PDZ domain containing family member 2 |
Gene Aliases: |
SEMCAP-2, SEMCAP2 |
Location: |
Chr.1:78044962-78138449 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GIPC2 | NM_001304725.1 | NP_001291654.1 | ||
| NM_017655.5 | 1 | 394 | NP_060125.4 | |
| AB073737.1 | 1 | 210 | BAB84711.1 | |
| AK225941.1 | 1 | 233 | ||
| AK307422.1 | ||||
| BC036075.1 | 1 | 394 | AAH36075.1 | |
| BI561167.1 | 1 | 400 |
More Information
Set Membership: |
|

Genomic Map