Assay Details
Target Gene Details
Entrez Gene ID: | 55831 |
Gene Name: | ER membrane protein complex subunit 3 |
Gene Aliases: |
POB, TMEM111 |
Location: |
Chr.3:9962680-10011248 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EMC3 | NM_018447.3 | NP_060917.1 | ||
| XM_005265321.3 | XP_005265378.1 | |||
| AF157321.1 | AAF67487.1 | |||
| AK172843.1 | BAD18807.1 | |||
| AK222953.1 | BAD96673.1 | |||
| AK312008.1 | ||||
| BC022807.1 | AAH22807.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv589648 | Chr.3:9892921 - 10042251 on Build GRCh38 | Loss |
|
| nsv834615 | Chr.3:9791053 - 9971639 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map