Assay Details
Target Gene Details
Entrez Gene ID: | 22916 |
Gene Name: | nuclear cap binding protein subunit 2 |
Gene Aliases: |
CBC2, CBP20, NIP1, PIG55 |
Location: |
Chr.3:196935402-196942594 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCBP2 | NM_001042540.1 | 4 | 624 | NP_001036005.1 |
| NM_001308036.1 | 4 | 1121 | NP_001294965.1 | |
| NM_007362.3 | 4 | 783 | NP_031388.2 | |
| XM_011512556.2 | 4 | 840 | XP_011510858.1 | |
| XM_011512557.2 | 4 | 888 | XP_011510859.1 | |
| XM_011512558.2 | 4 | 730 | XP_011510860.1 | |
| AK093216.1 | 4 | 833 | ||
| AK296144.1 | 4 | 635 | ||
| AK297506.1 | ||||
| AK308192.1 | 4 | 787 | ||
| AK315903.1 | 4 | 757 | ||
| BC001255.1 | 4 | 719 | AAH01255.1 | |
| BX537372.1 | 2 | 2780 | CAD97614.1 | |
| BX647994.1 | 4 | 768 | ||
| CN421262.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3599293 | Chr.3:196759100 - 197444213 on Build GRCh38 | Gain |
|
| nsv593086 | Chr.3:196897131 - 196958250 on Build GRCh38 | Loss |
|
| nsv1003135 | Chr.3:196621850 - 197012082 on Build GRCh38 | Gain |
|
| nsv819558 | Chr.3:196936635 - 196936996 on Build GRCh38 | Loss |
|
| esv2763775 | Chr.3:196727069 - 197015419 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map