Assay Details
Target Gene Details
Entrez Gene ID: | 8754 |
Gene Name: | ADAM metallopeptidase domain 9 |
Gene Aliases: |
CORD9, MCMP, MDC9, Mltng |
Location: |
Chr.8:38996771-39105261 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAM9 | NM_003816.2 | 1 | 48 | NP_003807.1 |
| NR_027638.1 | 1 | 48 | ||
| NR_027639.1 | 1 | 48 | ||
| NR_027878.1 | 1 | 48 | ||
| AF495383.1 | AAM49575.1 | |||
| BC126406.1 | ||||
| BC143923.1 | ||||
| U41766.1 | 1 | 48 | AAC50403.1 |
Target Gene Details
Entrez Gene ID: | 83877 |
Gene Name: | TM2 domain containing 2 |
Gene Aliases: |
BLP1 |
Location: |
Chr.8:38988808-38997139 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TM2D2 | XM_011544667.2 | 1 | 107 | XP_011542969.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv515080 | Chr.8:38996676 - 38997072 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map