Assay Details
Target Gene Details
Entrez Gene ID: | 55745 |
Gene Name: | adaptor related protein complex 5 mu 1 subunit |
Gene Aliases: |
C14orf108, MUDENG, Mu5, MuD |
Location: |
Chr.14:57268888-57298742 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP5M1 | NM_018229.3 | 2 | 930 | NP_060699.3 |
| NR_026895.1 | ||||
| XM_006720196.2 | 2 | 559 | XP_006720259.1 | |
| XM_011536940.2 | 3 | 902 | XP_011535242.1 | |
| AK001675.1 | 2 | 775 | BAA91828.1 | |
| AK172824.1 | BAD18791.1 | |||
| AK225345.1 | 2 | 777 | ||
| AK303999.1 | 3 | 804 | ||
| AL136685.1 | 2 | 909 | CAB66620.1 | |
| BC013174.1 | 2 | 894 | AAH13174.1 | |
| DB204683.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046422 | Chr.14:57198718 - 58129812 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map