Assay Details
Target Gene Details
Entrez Gene ID: | 83874 |
Gene Name: | TBC1 domain family member 10A |
Gene Aliases: |
EPI64, TBC1D10, dJ130H16.1, dJ130H16.2 |
Location: |
Chr.22:30291990-30326966 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D10A | NM_001204240.1 | 9 | 1462 | NP_001191169.1 |
| NM_031937.2 | 9 | 1441 | NP_114143.1 | |
| AB449894.1 | 9 | 1356 | ||
| AF331038.1 | 9 | 1380 | AAK35048.1 | |
| AK074656.1 | 9 | 1407 | BAC11117.1 | |
| AK093231.1 | 6 | 2576 | ||
| AK127939.1 | 9 | 1397 | ||
| AK131086.1 | 5 | 5006 | BAC85136.1 | |
| AK309455.1 | 9 | 1207 | ||
| BC136815.1 | 9 | 1397 | ||
| BC142940.1 | 9 | 1413 | ||
| BC150187.1 | 9 | 1361 | ||
| BC150214.1 | 9 | 1366 | ||
| BC171775.1 | 9 | 1397 | ||
| CT841514.1 | 9 | 1362 | CAJ86444.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv588885 | Chr.22:30265195 - 30295642 on Build GRCh38 | Loss |
|
| nsv522605 | Chr.22:30289087 - 30295642 on Build GRCh38 | Loss |
|
| nsv1062388 | Chr.22:30253190 - 30315454 on Build GRCh38 | Gain |
|
| nsv834174 | Chr.22:30223771 - 30350926 on Build GRCh38 | Gain+Loss |
|
| nsv829171 | Chr.22:30202041 - 30647591 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map