Assay Details
Target Gene Details
Entrez Gene ID: | 1583 |
Gene Name: | cytochrome P450 family 11 subfamily A member 1 |
Gene Aliases: |
CYP11A, CYPXIA1, P450SCC |
Location: |
Chr.15:74337762-74367740 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP11A1 | NM_000781.2 | NP_000772.2 | ||
| NM_001099773.1 | NP_001093243.1 | |||
| AK056794.1 | ||||
| AK225715.1 | ||||
| AK292300.1 | ||||
| AK292301.1 | ||||
| AK293204.1 | ||||
| AK293211.1 | ||||
| AK300178.1 | ||||
| AY603498.1 | ||||
| BC032329.1 | AAH32329.1 | |||
| BX398579.2 | ||||
| KU178011.1 | ||||
| KU178012.1 | ||||
| KU178013.1 | ||||
| KU178014.1 | ||||
| M14565.1 | AAA52162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map