Assay Details
Target Gene Details
Entrez Gene ID: | 79147 |
Gene Name: | fukutin related protein |
Gene Aliases: |
LGMD2I, MDC1C, MDDGA5, MDDGB5, MDDGC5 |
Location: |
Chr.19:46746015-46758575 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FKRP | NM_001039885.2 | 4 | 518 | NP_001034974.1 |
| NM_024301.4 | 4 | 466 | NP_077277.1 | |
| XM_005259247.2 | 5 | 579 | XP_005259304.1 | |
| XM_005259248.2 | 5 | 569 | XP_005259305.1 | |
| XM_005259249.4 | 5 | 511 | XP_005259306.1 | |
| XM_011527306.1 | 3 | 315 | XP_011525608.1 | |
| XM_011527307.1 | 4 | 364 | XP_011525609.1 | |
| XM_017027297.1 | 4 | 685 | XP_016882786.1 | |
| AJ314847.1 | 4 | 454 | CAC85633.1 | |
| AK022638.1 | 4 | 454 | BAB14146.1 | |
| AK095497.1 | 4 | 505 | ||
| AK291282.1 | 4 | 454 | ||
| BC002612.2 | 2 | 262 | AAH02612.1 | |
| BM979509.1 | ||||
| DA123579.1 | 4 | 395 | ||
| DN999716.1 | 1 | 187 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833850 | Chr.19:46703285 - 46867053 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map