Assay Details
Target Gene Details
Entrez Gene ID: | 4686 |
Gene Name: | nuclear cap binding protein subunit 1 |
Gene Aliases: |
CBP80, NCBP, Sto1 |
Location: |
Chr.9:97633423-97673748 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCBP1 | NM_002486.4 | 22 | 2657 | NP_002477.1 |
| XM_017014735.1 | 22 | 2495 | XP_016870224.1 | |
| XM_017014736.1 | 22 | 2208 | XP_016870225.1 | |
| XM_017014737.1 | 22 | 2196 | XP_016870226.1 | |
| XM_017014738.1 | 21 | 2196 | XP_016870227.1 | |
| AB209233.1 | 22 | 2237 | BAD92470.1 | |
| AK312807.1 | 22 | 2657 | ||
| BC001450.2 | 22 | 2261 | AAH01450.1 | |
| D32002.1 | 22 | 2462 | BAA06769.1 | |
| X80030.1 | 22 | 2228 | CAA56334.1 |
Target Gene Details
Entrez Gene ID: | 7507 |
Gene Name: | XPA, DNA damage recognition and repair factor |
Gene Aliases: |
XP1, XPAC |
Location: |
Chr.9:97654398-97697409 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XPA | XM_006717278.1 | XP_006717341.1 |
More Information
Set Membership: |
|

Genomic Map