Assay Details
Target Gene Details
Entrez Gene ID: | 390199 |
Gene Name: | olfactory receptor family 4 subfamily D member 9 |
Gene Aliases: |
OR11-253 |
Location: |
Chr.11:59514913-59515857 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR4D9 | NM_001004711.1 | 1 | 241 | NP_001004711.1 |
More Information
Set Membership: |
|

Genomic Map