Assay Details
Target Gene Details
Entrez Gene ID: | 3208 |
Gene Name: | hippocalcin |
Gene Aliases: |
BDR2, DYT2 |
Location: |
Chr.1:32885965-32894646 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HPCA | NM_002143.2 | 4 | 1096 | NP_002134.2 |
| BC001777.1 | 4 | 1096 | AAH01777.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950584 | Chr.1:32867900 - 32907199 on Build GRCh38 | Deletion |
|
| nsv516302 | Chr.1:32855000 - 32914507 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map