Assay Details
Target Gene Details
Entrez Gene ID: | 136371 |
Gene Name: | ankyrin repeat and SOCS box containing 10 |
Gene Aliases: |
GLC1F |
Location: |
Chr.7:151175698-151187832 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB10 | NM_001142459.1 | 1 | 353 | NP_001135931.2 |
| NM_001142460.1 | 1 | 353 | NP_001135932.2 | |
| NM_080871.3 | NP_543147.2 | |||
| AF417920.1 | 1 | 353 | AAL59159.1 | |
| AK055536.1 | 1 | 63 | ||
| AK125565.1 | BAC86204.1 | |||
| AK315114.1 | 1 | 181 | ||
| BC126351.1 | 1 | 117 | ||
| DA898875.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831188 | Chr.7:151038117 - 151215789 on Build GRCh38 | Loss |
|
| nsv609034 | Chr.7:151132928 - 151196543 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map