Assay Details
Target Gene Details
Entrez Gene ID: | 9338 |
Gene Name: | transcription elongation factor A like 1 |
Gene Aliases: |
SIIR, WEX9, p21, pp21 |
Location: |
Chr.X:103628720-103630953 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCEAL1 | NM_001006639.1 | NP_001006640.1 | ||
| NM_001006640.1 | NP_001006641.1 | |||
| NM_004780.2 | NP_004771.2 | |||
| BC000809.1 | AAH00809.1 | |||
| BM784458.1 | ||||
| BU602495.1 | ||||
| BX092470.1 | ||||
| CB053221.1 | ||||
| CF454849.1 | ||||
| M99701.1 | AAA60149.1 |
More Information
Set Membership: |
|

Genomic Map