Assay Details
Target Gene Details
Entrez Gene ID: | 50717 |
Gene Name: | DDB1 and CUL4 associated factor 8 |
Gene Aliases: |
GAN2, H326, WDR42A |
Location: |
Chr.1:160215715-160262560 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCAF8 | NM_015726.3 | 14 | 3829 | NP_056541.2 |
| NR_028103.1 | 14 | 4129 | ||
| NR_028104.1 | 13 | 4055 | ||
| AI352273.1 | 1 | 60 | ||
| BU629460.1 | 1 | 73 | ||
| U06631.1 | AAA16607.1 |
More Information
Set Membership: |
|

Genomic Map