Assay Details
Target Gene Details
Entrez Gene ID: | 97 |
Gene Name: | acylphosphatase 1 |
Gene Aliases: |
ACYPE |
Location: |
Chr.14:75053221-75069483 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACYP1 | NM_001107.4 | NP_001098.1 | ||
| NM_001302616.1 | NP_001289545.1 | |||
| NM_001302617.1 | NP_001289546.1 | |||
| NR_126393.1 | ||||
| NR_126394.1 | ||||
| NR_126395.1 | ||||
| AK312101.1 | ||||
| AW390293.1 | ||||
| BC035568.1 | AAH35568.1 | |||
| BG614847.1 | ||||
| BP316920.1 | ||||
| CD675334.1 | ||||
| DB071115.1 | ||||
| HY012032.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73309750] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map