Assay Details
Target Gene Details
Entrez Gene ID: | 404266 |
Gene Name: | HOXB cluster antisense RNA 3 |
Gene Aliases: |
- |
Location: |
Chr.17:48590420-48606414 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOXB-AS3 | NR_033201.2 | |||
| NR_033202.2 | ||||
| NR_033203.1 | ||||
| NR_033204.2 | ||||
| NR_110329.1 | ||||
| NR_110331.1 | ||||
| AA299267.1 | ||||
| BF431268.1 | ||||
| CB046509.1 | ||||
| CF127739.1 | ||||
| DA741273.1 | ||||
| W39335.1 |
Target Gene Details
Entrez Gene ID: | 3215 |
Gene Name: | homeobox B5 |
Gene Aliases: |
HHO.C10, HOX2, HOX2A, HU-1, Hox2.1 |
Location: |
Chr.17:48591257-48593741 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOXB5 | NM_002147.3 | 2 | 828 | NP_002138.1 |
| AK314964.1 | 2 | 885 | ||
| BC117247.1 | 2 | 789 | ||
| BE621135.1 | ||||
| M86726.1 | 1 | 122 | AAB59430.1 | |
| M92299.1 | 2 | 1044 | AAA52682.1 | |
| X03794.1 | 1 | 172 | CAA27420.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833471 | Chr.17:48540425 - 48705983 on Build GRCh38 | Gain |
|
| dgv5676n54 | Chr.17:48539657 - 48627174 on Build GRCh38 | Loss |
|
| nsv833469 | Chr.17:48414487 - 48632044 on Build GRCh38 | Loss |
|
| esv2758694 | Chr.17:48525311 - 48759475 on Build GRCh38 | Gain |
|
| dgv369n27 | Chr.17:48543040 - 48621348 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map