Assay Details
Target Gene Details
Entrez Gene ID: | 59277 |
Gene Name: | netrin 4 |
Gene Aliases: |
PRO3091 |
Location: |
Chr.12:95657807-95790758 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTN4 | NM_021229.3 | 10 | 2890 | NP_067052.2 |
| AF119916.3 | 11 | 2427 | AAF69670.2 | |
| AF278532.1 | 10 | 2892 | AAG30822.1 | |
| AF297711.1 | 10 | 2742 | AAG53651.1 | |
| AK024691.1 | 9 | 1966 | BAB14964.1 | |
| AK026833.1 | 5 | 1236 | ||
| AL137540.1 | 8 | 1758 | CAB70800.3 | |
| AL833767.1 | 9 | 2294 | CAH56243.1 | |
| BC013591.1 | 9 | 1965 | AAH13591.2 | |
| BC136798.1 | 10 | 2865 | ||
| BC136799.1 | 10 | 2865 |
Target Gene Details
Entrez Gene ID: | 100132594 |
Gene Name: | phosphoglycerate mutase 1 pseudogene 5 |
Gene Aliases: |
- |
Location: |
Chr.12:95649255-95673999 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGAM1P5 | NR_077225.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv455692 | Chr.12:95627257 - 95680547 on Build GRCh38 | Gain |
|
| nsv1052350 | Chr.12:95528912 - 95686094 on Build GRCh38 | Gain |
|
| nsv526022 | Chr.12:95658107 - 95665352 on Build GRCh38 | Loss |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map