Assay Details
Target Gene Details
Entrez Gene ID: | 9337 |
Gene Name: | CCR4-NOT transcription complex subunit 8 |
Gene Aliases: |
CAF1, CALIF, Caf1b, POP2, hCAF1 |
Location: |
Chr.5:154857952-154876792 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830521 | Chr.5:154768007 - 154864706 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map