Assay Details
Target Gene Details
Entrez Gene ID: | 84902 |
Gene Name: | centrosomal protein 89 |
Gene Aliases: |
CCDC123, CEP123 |
Location: |
Chr.19:32875925-32972029 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEP89 | NM_032816.4 | 19 | 2435 | NP_116205.3 |
| XM_005259344.3 | 19 | 2358 | XP_005259401.1 | |
| XM_011527425.2 | 16 | 1882 | XP_011525727.1 | |
| AK027546.1 | 18 | 2136 | BAB55190.1 | |
| AL832158.1 | 18 | 2258 | ||
| BC136328.1 | 19 | 2350 | ||
| BU628988.1 | 1 | 253 | ||
| DB539713.2 | 4 | 456 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1062926 | Chr.19:32828718 - 32904288 on Build GRCh38 | Loss |
|
| nsv516922 | Chr.19:32844072 - 32883216 on Build GRCh38 | Loss |
|
| nsv1060014 | Chr.19:32854920 - 33011720 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map