Assay Details
Target Gene Details
Entrez Gene ID: | 23023 |
Gene Name: | transmembrane and coiled-coil domain family 1 |
Gene Aliases: |
- |
Location: |
Chr.3:129647792-129893605 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 16 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508953 | Chr.3:129545350 - 129647876 on Build GRCh38 | Insertion |
|

Genomic Map