Assay Details
Target Gene Details
Entrez Gene ID: | 29763 |
Gene Name: | protein kinase C and casein kinase substrate in neurons 3 |
Gene Aliases: |
SDPIII |
Location: |
Chr.11:47177522-47186459 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PACSIN3 | NM_001184974.1 | 11 | 1818 | NP_001171903.1 |
| NM_001184975.1 | 11 | 1951 | NP_001171904.1 | |
| NM_016223.4 | 11 | 1822 | NP_057307.2 | |
| AF130979.1 | 11 | 1823 | AAF04472.1 | |
| AF149825.1 | 11 | 1759 | AAG31023.1 | |
| AF242530.1 | 11 | 1758 | AAK29207.1 | |
| BC007914.2 | 11 | 1751 | AAH07914.1 | |
| BC011889.2 | 11 | 1770 | AAH11889.1 |
More Information
Set Membership: |
|

Genomic Map