Assay Details
Target Gene Details
Entrez Gene ID: | 93589 |
Gene Name: | calcium voltage-gated channel auxiliary subunit alpha2delta 4 |
Gene Aliases: |
RCD4 |
Location: |
Chr.12:1791957-1922606 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 32 - Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNA2D4 | NM_172364.4 | NP_758952.4 | ||
| XM_011521041.2 | XP_011519343.1 | |||
| AF516695.1 | AAN06672.1 | |||
| AK301234.1 | ||||
| AL137658.1 | ||||
| BC048288.1 | AAH48288.1 | |||
| BC150186.1 | ||||
| BI908421.1 | ||||
| BX537436.1 | 32 | 3459 | CAD97678.1 | |
| BX537437.1 | CAD97679.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832305 | Chr.12:1660686 - 1826888 on Build GRCh38 | Gain |
|
| nsv556996 | Chr.12:1782206 - 1825754 on Build GRCh38 | Gain |
|
| dgv2273n54 | Chr.12:1798687 - 1824387 on Build GRCh38 | Loss |
|
| nsv832306 | Chr.12:1738560 - 1892142 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map