Assay Details
Target Gene Details
Entrez Gene ID: | 11066 |
Gene Name: | small nuclear ribonucleoprotein U11/U12 subunit 35 |
Gene Aliases: |
HM-1, U1SNRNPBP |
Location: |
Chr.12:123457644-123472362 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNRNP35 | NM_022717.3 | NP_073208.1 | ||
| NR_104103.1 | ||||
| XM_011537780.2 | XP_011536082.1 | |||
| XM_017018724.1 | XP_016874213.1 | |||
| AK290127.1 | ||||
| BC047678.1 | AAH47678.1 | |||
| BC054034.1 | AAH54034.1 | |||
| BM714052.1 | ||||
| U44799.1 | AAA86655.1 |
More Information
Set Membership: |
|

Genomic Map