Assay Details
Target Gene Details
Entrez Gene ID: | 5407 |
Gene Name: | pancreatic lipase related protein 1 |
Gene Aliases: |
PLRP1 |
Location: |
Chr.10:116590415-116609175 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNLIPRP1 | NM_001303135.1 | 7 | 646 | NP_001290064.1 |
| NM_006229.3 | 7 | 656 | NP_006220.1 | |
| XM_011539868.1 | 7 | 789 | XP_011538170.1 | |
| XM_011539869.1 | 7 | 1057 | XP_011538171.1 | |
| AK130236.1 | 7 | 644 | ||
| BC025784.2 | 6 | 619 | AAH25784.1 | |
| CR749299.1 | 6 | 2415 | CAH18154.1 | |
| CR749524.1 | 6 | 2415 | CAH18337.1 | |
| CR749596.1 | 6 | 2420 | CAH18394.1 | |
| M93283.1 | 7 | 638 | AAA59532.1 |
More Information
Set Membership: |
|

Genomic Map