Assay Details
Target Gene Details
Entrez Gene ID: | 8915 |
Gene Name: | B-cell CLL/lymphoma 10 |
Gene Aliases: |
CARMEN, CIPER, CLAP, IMD37, c-E10, mE10 |
Location: |
Chr.1:85265776-85276904 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCL10 | NM_001320715.1 | 2 | 781 | NP_001307644.1 |
| NM_003921.4 | 2 | 781 | NP_003912.1 | |
| XM_011542397.2 | 2 | 650 | XP_011540699.1 | |
| XM_011542398.2 | 2 | 655 | XP_011540700.1 | |
| XM_011542399.2 | 2 | 803 | XP_011540701.1 | |
| AF057700.1 | 2 | 475 | AAD15800.1 | |
| AF082283.1 | 3 | 935 | AAC99767.1 | |
| AF100338.1 | 2 | 476 | AAD16428.1 | |
| AF105066.1 | 2 | 229 | AAF06894.1 | |
| AF127386.1 | 2 | 229 | AAD32597.1 | |
| AF134395.1 | 2 | 474 | AAD39147.1 | |
| AJ006288.1 | 3 | 919 | CAA06955.1 | |
| AK291346.1 | 2 | 439 | ||
| BC053617.1 | 2 | 351 | AAH53617.1 | |
| BI831624.1 | ||||
| BX647306.1 | ||||
| EF189176.1 | 2 | 334 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34165 | Chr.1:85076357 - 85380566 on Build GRCh38 | Loss |
|
| nsv830459 | Chr.1:85123902 - 85320628 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map