Assay Details
Target Gene Details
Entrez Gene ID: | 4034 |
Gene Name: | leucine rich repeats and calponin homology domain containing 4 |
Gene Aliases: |
LRN, LRRN1, LRRN4, PP14183 |
Location: |
Chr.7:100574011-100586188 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRCH4 | NM_001289934.1 | NP_001276863.1 | ||
| NM_002319.4 | 18 | 2667 | NP_002310.2 | |
| AI491709.1 | ||||
| BC007634.1 | 17 | 2271 | AAH07634.1 |
Target Gene Details
Entrez Gene ID: | 101927655 |
Gene Name: | ZO-2 associated speckle protein |
Gene Aliases: |
- |
Location: |
Chr.7:100574035-100574733 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZASP | NM_001289933.1 | 1 | 154 | NP_001276862.1 |
| FR717257.1 | 1 | 154 | ||
| HQ401361.1 | 1 | 154 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
|
| nsv951386 | Chr.7:100566478 - 100682977 on Build GRCh38 | Deletion |
|
| nsv831072 | Chr.7:100564880 - 100765212 on Build GRCh38 | Loss |
|
| nsv5868 | Chr.7:100572375 - 100617023 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs144874653] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map