Assay Details
Target Gene Details
Entrez Gene ID: | 55177 |
Gene Name: | regulator of microtubule dynamics 3 |
Gene Aliases: |
FAM82A2, FAM82C, RMD-3, RMD3, ptpip51 |
Location: |
Chr.15:40735884-40755336 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RMDN3 | NM_001304802.1 | 13 | 1931 | NP_001291731.1 |
| NM_001323894.1 | 13 | 1845 | NP_001310823.1 | |
| NM_001323895.1 | 13 | 2062 | NP_001310824.1 | |
| NM_001323896.1 | 13 | 1923 | NP_001310825.1 | |
| NM_001323897.1 | 13 | 2039 | NP_001310826.1 | |
| NM_018145.2 | 13 | 1961 | NP_060615.1 | |
| XM_011521755.2 | 10 | 2207 | XP_011520057.1 | |
| AA772280.1 | 1 | 377 | ||
| AK001441.1 | 13 | 1885 | BAA91693.1 | |
| AK025963.1 | 10 | 1274 | BAB15298.1 | |
| AK090731.1 | 6 | 1833 | ||
| AK092058.1 | 13 | 1844 | ||
| AK097286.1 | 14 | 1773 | ||
| AK123192.1 | 14 | 1838 | BAC85554.1 | |
| AK123282.1 | 12 | 2768 | ||
| AY358793.1 | 12 | 1701 | AAQ89153.1 | |
| BC008970.2 | 9 | 1158 | AAH08970.2 | |
| BC063844.1 | 13 | 1849 | AAH63844.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527234 | Chr.15:40695527 - 40739488 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map