Assay Details
Target Gene Details
Entrez Gene ID: | 23357 |
Gene Name: | angel homolog 1 |
Gene Aliases: |
Ccr4e, KIAA0759 |
Location: |
Chr.14:76786473-76812947 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANGEL1 | NM_015305.3 | 10 | 3882 | NP_056120.2 |
| XM_017021115.1 | XP_016876604.1 | |||
| AB018302.1 | 10 | 3779 | ||
| AF275805.1 | 1 | 252 | ||
| AK025609.1 | 1 | 1808 | ||
| AL137268.1 | 10 | 3780 | CAB70667.2 | |
| BC028714.2 | 10 | 3829 | AAH28714.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952869 | Chr.14:76787458 - 76790357 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map