Assay Details
Target Gene Details
Entrez Gene ID: | 11066 |
Gene Name: | small nuclear ribonucleoprotein U11/U12 subunit 35 |
Gene Aliases: |
HM-1, U1SNRNPBP |
Location: |
Chr.12:123457644-123472362 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNRNP35 | NM_022717.3 | 2 | 165 | NP_073208.1 |
| NM_180699.3 | 2 | 249 | NP_851030.1 | |
| NR_104103.1 | ||||
| XM_006719204.3 | 2 | 1348 | XP_006719267.1 | |
| XM_011537780.2 | 2 | 625 | XP_011536082.1 | |
| XM_017018724.1 | 3 | 1076 | XP_016874213.1 | |
| AK290127.1 | 2 | 145 | ||
| BC009622.1 | 3 | 575 | AAH09622.1 | |
| BC047678.1 | 6 | 670 | AAH47678.1 | |
| BC054034.1 | 2 | 82 | AAH54034.1 | |
| BC093671.1 | 2 | 114 | AAH93671.1 | |
| BC111955.1 | 2 | 114 | AAI11956.1 | |
| BM714052.1 | 2 | 165 | ||
| U44798.1 | 3 | 262 | AAA86654.1 | |
| U44799.1 | 2 | 109 | AAA86655.1 |
More Information
Set Membership: |
|

Genomic Map