Assay Details
Target Gene Details
Entrez Gene ID: | 1819 |
Gene Name: | developmentally regulated GTP binding protein 2 |
Gene Aliases: |
- |
Location: |
Chr.17:18087867-18107985 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DRG2 | NM_001388.4 | NP_001379.1 | ||
| XM_005256499.2 | XP_005256556.1 | |||
| XM_005256500.2 | XP_005256557.1 | |||
| XM_011523704.1 | XP_011522006.1 | |||
| XM_011523705.1 | XP_011522007.1 | |||
| XM_011523706.1 | XP_011522008.1 | |||
| XM_011523707.1 | XP_011522009.1 | |||
| XM_017024296.1 | XP_016879785.1 | |||
| XM_017024297.1 | XP_016879786.1 | |||
| AB209340.1 | BAD92577.1 | |||
| AK297569.1 | ||||
| AK313362.1 | ||||
| BC000493.2 | AAH00493.1 | |||
| BT006976.1 | AAP35622.1 | |||
| X80754.1 | CAA56730.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574527 | Chr.17:18100531 - 18142976 on Build GRCh38 | Loss |
|
| nsv953839 | Chr.17:18089487 - 18146586 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map