Assay Details
Target Gene Details
Entrez Gene ID: | 5298 |
Gene Name: | phosphatidylinositol 4-kinase beta |
Gene Aliases: |
NPIK, PI4K-BETA, PI4K92, PI4KBETA, PI4KIIIBETA, PIK4CB |
Location: |
Chr.1:151291797-151327715 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PI4KB | NM_001198773.1 | 4 | 1628 | NP_001185702.1 |
| NM_001198774.1 | 2 | 1311 | NP_001185703.1 | |
| NM_001198775.1 | NP_001185704.1 | |||
| NM_002651.2 | 3 | 1455 | NP_002642.1 | |
| XM_005245264.3 | 3 | 1455 | XP_005245321.1 | |
| XM_011509634.1 | 3 | 1455 | XP_011507936.1 | |
| XM_011509635.1 | 4 | 1628 | XP_011507937.1 | |
| XM_011509636.2 | 2 | 1283 | XP_011507938.1 | |
| XM_011509637.2 | 3 | 1456 | XP_011507939.1 | |
| XM_011509638.1 | 3 | 1455 | XP_011507940.1 | |
| XM_017001486.1 | 4 | 1628 | XP_016856975.1 | |
| XM_017001487.1 | 2 | 1283 | XP_016856976.1 | |
| XM_017001488.1 | 3 | 1456 | XP_016856977.1 | |
| XM_017001489.1 | 4 | 1628 | XP_016856978.1 | |
| XM_017001490.1 | 2 | 1283 | XP_016856979.1 | |
| XM_017001491.1 | 3 | 1456 | XP_016856980.1 | |
| XM_017001492.1 | 3 | 1456 | XP_016856981.1 | |
| AB005910.1 | 2 | 943 | BAA21661.1 | |
| AJ011121.1 | 2 | 928 | CAA09495.1 | |
| AK294606.1 | ||||
| AK311292.1 | ||||
| BC000029.2 | 2 | 991 | AAH00029.1 | |
| BC040300.1 | 2 | 1311 | AAH40300.1 | |
| DC350175.1 | ||||
| DC365634.1 | ||||
| U81802.1 | 3 | 1127 | AAC51156.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522840 | Chr.1:150961247 - 151375270 on Build GRCh38 | Gain |
|
| esv34461 | Chr.1:151268066 - 151439159 on Build GRCh38 | Gain |
|
| nsv2821 | Chr.1:151298648 - 151329483 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map