Assay Details
Target Gene Details
Entrez Gene ID: | 148418 |
Gene Name: | sterile alpha motif domain containing 13 |
Gene Aliases: |
HSD-41, HSD-42 |
Location: |
Chr.1:84298366-84350798 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAMD13 | NM_001010971.2 | NP_001010971.1 | ||
| NM_001134663.1 | NP_001128135.1 | |||
| NM_001134664.1 | NP_001128136.1 | |||
| XM_017000377.1 | XP_016855866.1 | |||
| AK056937.1 | ||||
| DB459148.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830426 | Chr.1:84239241 - 84390546 on Build GRCh38 | Gain |
|
| nsv830414 | Chr.1:84171055 - 84358968 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map