Assay Details
Target Gene Details
Entrez Gene ID: | 4128 |
Gene Name: | monoamine oxidase A |
Gene Aliases: |
MAO-A |
Location: |
Chr.X:43654907-43746824 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAOA | NM_000240.3 | 3 | 1659 | NP_000231.1 |
| NM_001270458.1 | 4 | 1767 | NP_001257387.1 | |
| AK223499.1 | 3 | 401 | BAD97219.1 | |
| AK291769.1 | 3 | 263 | ||
| AK293926.1 | 4 | 374 | ||
| BC008064.2 | 3 | 245 | AAH08064.1 | |
| BC044787.1 | 3 | 1661 | AAH44787.1 | |
| BT006651.1 | 3 | 224 | AAP35297.1 | |
| BX501303.1 | 3 | 415 | ||
| M68840.1 | 3 | 297 | AAA59548.1 | |
| M69226.1 | 3 | 274 | AAA59549.1 |
More Information
Set Membership: |
|

Genomic Map