Assay Details
Target Gene Details
Entrez Gene ID: | 25841 |
Gene Name: | ankyrin repeat and BTB domain containing 2 |
Gene Aliases: |
ABTB2A, BTBD22 |
Location: |
Chr.11:34150987-34358008 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABTB2 | NM_145804.2 | NP_665803.2 | ||
| AK091560.1 | BAC03697.1 | |||
| AK095632.1 | BAC04595.1 | |||
| AK291744.1 | ||||
| BC093968.1 | AAH93968.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509400 | Chr.11:34110735 - 34232487 on Build GRCh38 | Insertion |
|
| nsv951600 | Chr.11:34136054 - 34174953 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map