Assay Details
Target Gene Details
Entrez Gene ID: | 89849 |
Gene Name: | autophagy related 16 like 2 |
Gene Aliases: |
ATG16B, WDR80 |
Location: |
Chr.11:72814406-72843740 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATG16L2 | NM_001318766.1 | 18 | 2009 | NP_001305695.1 |
| NM_033388.1 | 18 | 1889 | NP_203746.1 | |
| XM_005274376.4 | XP_005274433.1 | |||
| XM_006718732.2 | 17 | 1825 | XP_006718795.1 | |
| XM_006718734.2 | 10 | 3068 | XP_006718797.1 | |
| XM_011545332.1 | 17 | 1774 | XP_011543634.1 | |
| XM_011545333.1 | 18 | 2337 | XP_011543635.1 | |
| XM_011545334.1 | 18 | 1750 | XP_011543636.1 | |
| AK024423.1 | 8 | 4323 | BAB15713.1 | |
| AK090597.1 | 18 | 2000 | BAC03485.1 | |
| AK091151.1 | 17 | 3477 | ||
| AK093017.1 | 18 | 1862 | BAC04021.1 | |
| AL832974.1 | 10 | 1500 | CAH56355.1 | |
| BC036713.2 | 16 | 1618 | AAH36713.1 | |
| BC137489.1 | 18 | 1987 | ||
| BC137490.1 | 18 | 1987 | ||
| BC142718.1 | 16 | 3585 | ||
| BC146660.1 | 16 | 3585 | ||
| BQ929430.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951039 | Chr.11:72816356 - 72832855 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map