Assay Details
Target Gene Details
Entrez Gene ID: | 6421 |
Gene Name: | splicing factor proline and glutamine rich |
Gene Aliases: |
POMP100, PPP1R140, PSF |
Location: |
Chr.1:35176378-35193161 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SFPQ | NM_005066.2 | 5 | 1589 | NP_005057.1 |
| NR_136702.1 | 5 | 1592 | ||
| NR_136703.1 | 5 | 1592 | ||
| XM_005271112.4 | 5 | 1608 | XP_005271169.1 | |
| XM_005271113.4 | 5 | 1608 | XP_005271170.1 | |
| XM_005271115.4 | 5 | 1608 | XP_005271172.1 | |
| XM_011541950.2 | 5 | 1608 | XP_011540252.1 | |
| XM_017002053.1 | 5 | 1608 | XP_016857542.1 | |
| XM_017002054.1 | 5 | 1608 | XP_016857543.1 | |
| BC004534.2 | 5 | 1277 | AAH04534.2 | |
| BC027708.2 | 5 | 1580 | AAH27708.1 | |
| BC051192.1 | 5 | 1591 | AAH51192.1 | |
| X16850.1 | 4 | 563 | CAA34747.1 | |
| X70944.1 | 5 | 1581 | CAA50283.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508149 | Chr.1:35187187 - 35285669 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map