Assay Details
Target Gene Details
Entrez Gene ID: | 6097 |
Gene Name: | RAR related orphan receptor C |
Gene Aliases: |
IMD42, NR1F3, RORG, RZR-GAMMA, RZRG, TOR |
Location: |
Chr.1:151806071-151832450 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RORC | NM_001001523.1 | NP_001001523.1 | ||
| NM_005060.3 | NP_005051.2 | |||
| XM_006711484.3 | XP_006711547.2 | |||
| AB307696.1 | ||||
| AK097994.1 | ||||
| AK128522.1 | ||||
| AK223137.1 | BAD96857.1 | |||
| AK298460.1 | ||||
| AK315662.1 | ||||
| AL834219.1 | CAD38900.1 | |||
| BC031554.1 | AAH31554.1 | |||
| BC110571.2 | AAI10572.1 | |||
| CR457436.1 | CAG33717.1 | |||
| HQ692834.1 | ||||
| U16997.1 | AAA64751.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516369 | Chr.1:151807701 - 151847848 on Build GRCh38 | Gain |
|
| esv3578175 | Chr.1:151807676 - 151811377 on Build GRCh38 | Loss |
|
| nsv1010398 | Chr.1:151789173 - 151818464 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map