Assay Details
Target Gene Details
Entrez Gene ID: | 10625 |
Gene Name: | influenza virus NS1A binding protein |
Gene Aliases: |
FLARA3, HSPC068, KLHL39, ND1, NS-1, NS1-BP, NS1BP |
Location: |
Chr.1:185292367-185317343 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IVNS1ABP | NM_006469.4 | 15 | 3567 | NP_006460.2 |
| XM_005244843.3 | 16 | 3664 | XP_005244900.1 | |
| XM_011509079.2 | 15 | 3598 | XP_011507381.1 | |
| XM_011509080.2 | 14 | 3432 | XP_011507382.1 | |
| XM_017000084.1 | 16 | 3685 | XP_016855573.1 | |
| AB020657.1 | 15 | 3574 | ||
| AF205218.1 | 15 | 3499 | AAG43485.1 | |
| AK000931.1 | 8 | 2207 | ||
| AK001625.1 | 13 | 2905 | ||
| AK001824.1 | 8 | 2285 | ||
| AK023123.1 | ||||
| AL137640.1 | 1 | 895 | ||
| BX648273.1 | 8 | 3144 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548398 | Chr.1:185167326 - 186033962 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map