Assay Details
Target Gene Details
Entrez Gene ID: | 1488 |
Gene Name: | C-terminal binding protein 2 |
Gene Aliases: |
- |
Location: |
Chr.10:124987849-125161061 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv51n21 | Chr.10:124995565 - 125004911 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map