Assay Details
Target Gene Details
Entrez Gene ID: | 2319 |
Gene Name: | flotillin 2 |
Gene Aliases: |
ECS-1, ECS1, ESA, ESA1, M17S1 |
Location: |
Chr.17:28879339-28897728 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FLOT2 | NM_004475.2 | 11 | 1453 | NP_004466.2 |
| XM_005257950.3 | 11 | 1484 | XP_005258007.1 | |
| XM_005257953.2 | 10 | 1388 | XP_005258010.1 | |
| XM_017024394.1 | 11 | 1381 | XP_016879883.1 | |
| XM_017024395.1 | 12 | 1550 | XP_016879884.1 | |
| XM_017024396.1 | 11 | 1387 | XP_016879885.1 | |
| AK225491.1 | 4 | 483 | ||
| BC003683.1 | AAH03683.1 | |||
| BC017292.1 | 10 | 1275 | ||
| BT019477.1 | AAV38284.1 | |||
| BT019478.1 | AAV38285.1 | |||
| CR541784.1 | CAG46583.1 | |||
| CR542265.1 | CAG47061.1 | |||
| M60922.1 | 11 | 1309 | AAA65729.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574662 | Chr.17:28867616 - 29014758 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map