Assay Details
Target Gene Details
Entrez Gene ID: | 132321 |
Gene Name: | chromosome 4 open reading frame 33 |
Gene Aliases: |
- |
Location: |
Chr.4:129093674-129112688 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C4orf33 | NM_173487.2 | NP_775758.2 | ||
| XM_017007724.1 | XP_016863213.1 | |||
| AK091022.1 | BAC03568.1 | |||
| AK309229.1 | ||||
| BC032582.1 | AAH32582.1 | |||
| BP871612.1 | ||||
| BQ549846.1 | ||||
| BX538164.1 | CAD98044.1 |
More Information
Set Membership: |
|

Genomic Map