Assay Details
Target Gene Details
Entrez Gene ID: | 3069 |
Gene Name: | high density lipoprotein binding protein |
Gene Aliases: |
HBP, PRO2900, VGL |
Location: |
Chr.2:241227264-241315842 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDLBP | NM_005336.5 | 1 | 200 | NP_005327.1 |
| XM_011511058.2 | 1 | 173 | XP_011509360.1 | |
| XM_011511060.2 | 1 | 173 | XP_011509362.1 | |
| XM_017003941.1 | 1 | 195 | XP_016859430.1 | |
| AK296684.1 | ||||
| AK312335.1 | ||||
| BC001179.1 | AAH01179.1 | |||
| BG720303.1 | 1 | 202 | ||
| BX647759.1 | ||||
| DC424146.1 | 1 | 200 |
Target Gene Details
Entrez Gene ID: | 4735 |
Gene Name: | septin 2 |
Gene Aliases: |
DIFF6, NEDD-5, NEDD5, Pnutl3, hNedd5 |
Location: |
Chr.2:241315187-241354027 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEPT2 | NM_001008491.2 | NP_001008491.1 | ||
| NM_006155.2 | NP_006146.1 | |||
| XM_017004195.1 | XP_016859684.1 | |||
| XM_017004201.1 | XP_016859690.1 | |||
| XM_017004202.1 | XP_016859691.1 | |||
| XM_017004212.1 | XP_016859701.1 | |||
| BC014455.1 | AAH14455.1 | |||
| BI561073.1 | ||||
| D63878.1 | ||||
| DB035463.1 | ||||
| DB095611.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3594825 | Chr.2:241310914 - 241318141 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77247061] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map