Assay Details
Target Gene Details
Entrez Gene ID: | 57504 |
Gene Name: | metastasis associated 1 family member 3 |
Gene Aliases: |
- |
Location: |
Chr.2:42494569-42795032 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTA3 |
Target Gene Details
Entrez Gene ID: | 165140 |
Gene Name: | oxoeicosanoid receptor 1 |
Gene Aliases: |
GPCR, GPR170, TG1019 |
Location: |
Chr.2:42762499-42764261 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OXER1 | NM_148962.4 | 1 | 136 | NP_683765.1 |
| AB083055.2 | 1 | 92 | BAC11806.2 | |
| BC063549.1 | 1 | 136 | AAH63549.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519877 | Chr.2:42751296 - 42793774 on Build GRCh38 | Loss |
|
| nsv457152 | Chr.2:42752929 - 42787149 on Build GRCh38 | Loss |
|
| nsv817795 | Chr.2:42751092 - 42768708 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map