Assay Details
Target Gene Details
Entrez Gene ID: | 2784 |
Gene Name: | G protein subunit beta 3 |
Gene Aliases: |
CSNB1H |
Location: |
Chr.12:6840854-6847400 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNB3 | NM_001297571.1 | 8 | 759 | NP_001284500.1 |
| NM_002075.3 | 8 | 762 | NP_002066.1 | |
| XM_011520953.2 | 8 | 694 | XP_011519255.1 | |
| AB209283.1 | 7 | 1903 | BAD92520.1 | |
| AB593098.1 | 8 | 688 | ||
| AB593099.1 | 8 | 687 | ||
| AF501884.1 | 7 | 552 | AAM15920.1 | |
| AK313091.1 | 8 | 639 | ||
| BC000115.1 | 8 | 675 | AAH00115.1 | |
| BC002454.2 | 8 | 682 | AAH02454.1 | |
| BC015920.1 | 8 | 641 | AAH15920.1 | |
| BT009800.1 | 7 | 552 | AAP88802.1 | |
| M31328.1 | 7 | 558 | AAA52582.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv557261 | Chr.12:6839304 - 6924660 on Build GRCh38 | Loss |
|
| nsv832324 | Chr.12:6780297 - 6932306 on Build GRCh38 | Loss |
|
| esv3580405 | Chr.12:6826466 - 6903969 on Build GRCh38 | Gain |
|
| nsv832323 | Chr.12:6662233 - 6856559 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map