Assay Details
Target Gene Details
Entrez Gene ID: | 140460 |
Gene Name: | ankyrin repeat and SOCS box containing 7 |
Gene Aliases: |
- |
Location: |
Chr.15:100602550-100651701 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB7 | NM_024708.3 | 1 | 344 | NP_078984.2 |
| NM_198243.2 | 1 | 344 | NP_937886.1 | |
| AF451994.1 | 1 | 264 | AAP97683.1 | |
| AK026204.1 | 1 | 264 | BAB15392.1 | |
| AK222958.1 | 1 | 264 | BAD96678.1 | |
| AK222974.1 | 1 | 264 | BAD96694.1 | |
| AK289860.1 | 1 | 297 | ||
| BC063581.1 | 1 | 285 | AAH63581.1 | |
| CD632230.1 | 1 | 348 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1041172 | Chr.15:100435008 - 101062634 on Build GRCh38 | Gain |
|
| nsv833107 | Chr.15:100474881 - 100650621 on Build GRCh38 | Gain |
|
| esv3637480 | Chr.15:100508357 - 100603781 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map