Assay Details
Target Gene Details
Entrez Gene ID: | 79005 |
Gene Name: | sodium channel modifier 1 |
Gene Aliases: |
- |
Location: |
Chr.1:151166022-151170297 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCNM1 | NM_001204856.1 | 7 | 840 | NP_001191785.1 |
| NM_024041.3 | 7 | 791 | NP_076946.1 | |
| NR_037937.1 | 7 | 741 | ||
| AA374943.1 | 1 | 88 | ||
| AK056322.1 | 7 | 760 | BAB71149.1 | |
| AV718162.2 | ||||
| BC000264.1 | 7 | 735 | AAH00264.1 | |
| BU540926.1 | 7 | 607 |
Target Gene Details
Entrez Gene ID: | 100534012 |
Gene Name: | TNFAIP8L2-SCNM1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.1:151156629-151170297 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFAIP8L2-SCNM1 | NM_001204848.1 | 7 | 703 | NP_001191777.1 |
| BG531092.1 | 7 | 670 | ||
| CR999387.1 | 6 | 684 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522840 | Chr.1:150961247 - 151375270 on Build GRCh38 | Gain |
|
| dgv2n68 | Chr.1:151136983 - 151309302 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114031237] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map