Assay Details
Target Gene Details
Entrez Gene ID: | 55731 |
Gene Name: | family with sequence similarity 222 member B |
Gene Aliases: |
C17orf63 |
Location: |
Chr.17:28755978-28855036 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entrez Gene ID: | 55731 |
Gene Name: | family with sequence similarity 222 member B |
Gene Aliases: |
C17orf63 |
Location: |
Chr.17:28755978-28855036 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |