Assay Details
Target Gene Details
Entrez Gene ID: | 1135 |
Gene Name: | cholinergic receptor nicotinic alpha 2 subunit |
Gene Aliases: |
- |
Location: |
Chr.8:27459761-27479296 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRNA2 | NM_000742.3 | 3 | 748 | NP_000733.2 |
| NM_001282455.1 | 3 | 748 | NP_001269384.1 | |
| XM_005273397.2 | 3 | 672 | XP_005273454.1 | |
| XM_006716282.1 | 4 | 659 | XP_006716345.1 | |
| XM_011544388.1 | 4 | 750 | XP_011542690.1 | |
| XM_011544389.1 | 3 | 748 | XP_011542691.1 | |
| XM_017013003.1 | 3 | 725 | XP_016868492.1 | |
| XM_017013004.1 | 4 | 659 | XP_016868493.1 | |
| AK296348.1 | 3 | 303 | ||
| AK314787.1 | 3 | 666 | ||
| BC153866.1 | 2 | 174 | ||
| CD013901.1 | 2 | 314 | ||
| DA096824.1 | 2 | 140 | ||
| U62431.1 | 3 | 693 | AAB40109.1 | |
| Y16281.1 | 2 | 139 | CAA76154.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1020582 | Chr.8:27452251 - 27549327 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map