Assay Details
Target Gene Details
Entrez Gene ID: | 374407 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member B13 |
Gene Aliases: |
RSPH16A, TSARG5, TSARG6 |
Location: |
Chr.11:73950319-73970287 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJB13 | NM_153614.3 | 1 | 540 | NP_705842.2 |
| XM_005273984.3 | 1 | 67 | XP_005274041.1 | |
| XM_011545004.2 | 1 | 232 | XP_011543306.1 | |
| XM_011545005.2 | 1 | 234 | XP_011543307.1 | |
| XM_011545009.2 | 1 | 65 | XP_011543311.1 | |
| XM_011545013.2 | 1 | 238 | XP_011543315.1 | |
| XM_011545014.2 | 1 | 238 | XP_011543316.1 | |
| XM_017017679.1 | 1 | 238 | XP_016873168.1 | |
| AY138810.2 | 1 | 540 | AAN15929.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73543095,rs73543099] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map