Assay Details
Target Gene Details
Entrez Gene ID: | 10962 |
Gene Name: | myeloid/lymphoid or mixed-lineage leukemia; translocated to, 11 |
Gene Aliases: |
AF1Q |
Location: |
Chr.1:151059675-151068497 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MLLT11 | NM_006818.3 | NP_006809.1 | ||
| AK056089.1 | ||||
| AK057755.1 | ||||
| BC006471.2 | ||||
| BC008445.1 | AAH08445.1 | |||
| BC009624.1 | AAH09624.1 | |||
| BC019253.1 | AAH19253.1 | |||
| BC021703.1 | AAH21703.1 | |||
| BC022448.1 | AAH22448.1 | |||
| BT006799.1 | AAP35445.1 | |||
| CR456879.1 | CAG33160.1 | |||
| CR542150.1 | CAG46947.1 | |||
| U16954.1 | AAA70088.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522840 | Chr.1:150961247 - 151375270 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map